2-(4-Methoxyphenyl)ethanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:702-23-8
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00002900
:152.19 g/mol
Boiling Pt:334…336 °C
Melting Pt:26…30 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:211-866-6
Related Information
Frequently Bought Together