
α-Bromo-p-toluic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6232-88-8
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00002567
:215.05 g/mol
Boiling Pt:316 °C (1013 hPa)
Melting Pt:223…227 °C
Storage Temperature:Ambient
EINECS:228-343-3
UN:3261
ADR:8,II
:GHS05
Related Information
Frequently Bought Together