Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Dimethylbenzenesulphonyl chloride ≥98%
Corrosive
2,4-Dimethylbenzenesulphonyl chloride ≥98%
Catalog # H50301.03
CAS Number:  
Corrosive
2,4-Dimethylbenzenesulphonyl chloride ≥98%
Catalog # H50301.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:609-60-9
 :
Formula:C₈H₉ClO₂S
MDL Number:MFCD00156143
 :204.68 g/mol
Melting Pt:32…33 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05