

4-Bromo-2-methylphenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19241-38-4
:
Formula:C₈H₆BrNS
MDL Number:MFCD00041087
:228.11 g/mol
Boiling Pt:129…130 °C (3 mmHg)
Melting Pt:44…46 °C
Storage Temperature:Ambient
EINECS:242-907-6
UN:2923
ADR:8,III
:GHS06
:GHS05
Frequently Bought Together