N-(6-Bromohexyl)phthalimide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24566-79-8
:
Formula:C₁₄H₁₆BrNO₂
MDL Number:MFCD00023098
:310.19 g/mol
Boiling Pt:186…188 °C (0.4 mmHg)
Melting Pt:54…60 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together