
3-(Trifluoromethyl)benzylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2740-83-2
:
Formula:C₈H₈F₃N
MDL Number:MFCD00008117
Boiling Pt:93…97 °C (28 mmHg)
Density:1.222
Flash Pt:71 °C (159 °F)
Storage Temperature:Ambient
EINECS:220-367-2
ADR:8,III
:175.15 g/mol
:GHS05
Frequently Bought Together