

2-Chloroethyl methyl ether ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:627-42-9
:
Formula:C₃H₇ClO
MDL Number:MFCD00000972
:94.54 g/mol
Boiling Pt:89…90 °C
Melting Pt:–55…55 °C
Density:1.04
Flash Pt:8 °C (46 °F)
Storage Temperature:Ambient
EINECS:210-999-7
UN:3271
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together