

Tetramethylammonium fluoride tetrahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17787-40-5
:
Formula:C₄H₁₂FN·4H₂O
MDL Number:MFCD00149968
:165.21 g/mol
Melting Pt:42…46 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:206-769-0
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together