


(Ethyl)triphenylphosphonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4736-60-1
:
Formula:C₂₀H₂₀IP
MDL Number:MFCD00040352
:418.26 g/mol
Melting Pt:164…169 °C
Density:1 g/cm³ (20 °C)
EINECS:225-245-2
UN:2811
ADR:6.1,III
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together