2-Chlorobenzyl mercaptan ≥97%
: 2-Chlorobenzenemethanethiol, o-Chlorotoluene-α-thiol
:39718-00-8
:
:C₇H₇ClS
:MFCD00004868
:158.65
:120…122 °C (25 mmHg)
:1.220
:69 °C (156 °F)
:Ambient
:254-602-5
:2810
:6.1,III
:GHS06
:GHS07
- Environmentally Preferable:
Frequently Bought Together