
Octadecyltrichlorosilane 95% cont. 5-10% branched isomers
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-04-9
:
Formula:C₁₈H₃₇Cl₃Si
MDL Number:MFCD00000484
:387.94 g/mol
Boiling Pt:160…162 °C (3 mmHg)
Melting Pt:∼ 20 °C
Density:0.984
Flash Pt:189 °C (372 °F)
Storage Temperature:Ambient
EINECS:203-930-7
UN:1800
ADR:8,II
:GHS05
Related Information
Frequently Bought Together