
Tris(4-fluorophenyl)phosphine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18437-78-0
:
Formula:C₁₈H₁₂F₃P
MDL Number:MFCD00013553
:316.26 g/mol
Melting Pt:78…82 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together