
3,5-Bis(trifluoromethyl)phenol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:349-58-6
:
Formula:C₈H₄F₆O
MDL Number:MFCD00000386
:230.11 g/mol
Boiling Pt:97…98 °C (50 mmHg)
Melting Pt:20…21 °C
Density:1.511
Flash Pt:>110 °C (230 °F)
EINECS:206-488-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together