
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6001-64-5
:
Formula:C₈H₁₆Cl₆O₃
MDL Number:MFCD02179352
:372.93 g/mol
Merck Index:14,2129
Boiling Pt:167 °C
Melting Pt:98…100 °C (anhydr.)
Flash Pt:>100 °C (212 °F)
EINECS:200-317-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together