

N,O-Bis(trimethylsilyl)trifluoroacetamide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25561-30-2
:
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269
:257.4 g/mol
Boiling Pt:145…147 °C
Melting Pt:–10 °C
Density:0.97
Flash Pt:23 °C (73 °F)
UN:2924
ADR:3,III
:Packaged under argon in resealable ChemSeal™ bottles.
EINECS:247-103-9
:GHS05
:GHS02
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together