

Disodium hexachloroiridate hexahydrate (≥33.9% Ir)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19567-78-3
:
Formula:Cl₆IrNa₂·6H₂O
MDL Number:MFCD00149169
:559.01 g/mol
Melting Pt:600 °C (dec.)
EINECS:241-011-2
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together