
4-Cyanophenyl isocyanate 95%
:40465-45-0
:
:C₈H₄N₂O
:144.13 g/mol
:123 °C (12 mmHg)
:102 °C
:Ambient
:254-934-0
:2206
:6.1,III
:GHS07
- Appearance:Off-white to beige Crystalline needles or solid
- Infrared spectrum:Conforms
- Melting point:99 to 106 °C
- GC:≥94 %
Frequently Bought Together