
2-(4-Aminophenyl)ethanol 97%
:104-10-9
:
:C₈H₁₁NO
:MFCD00007922
:137.18 g/mol
:235 - 255 °C (1013 hPa)
:104…108 °C
:Ambient
:203-174-8
:6.1,III
:GHS07
- Appearance:Off-white to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:106 to 110 °C
- GC:≥96.0 %
Frequently Bought Together