

α-Bromo-4-nitro-o-cresol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:772-33-8
:
Formula:C₇H₆BrNO₃
MDL Number:MFCD00007340
:232.03
Melting Pt:145…149 °C
Density:Min, 1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-248-9
UN:2923
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:Beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:143 to 149 °C
- Titration Argentometric:≥94 %
Specification Test Results
Frequently Bought Together