Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Methylphenyl)piperazine 98%
Irritant
Corrosive
1-(2-Methylphenyl)piperazine 98%
Certificates
undefined
Irritant
Corrosive
1-(2-Methylphenyl)piperazine 98%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals or crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    43 to 48 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together