ContactUsLinkComponent
n-Butyllithium 2.7 M in toluene, AcroSeal®
Irritant
Health
Corrosive
Flammable
n-Butyllithium 2.7 M in toluene, AcroSeal®
Certificates
undefined
Irritant
Health
Corrosive
Flammable
n-Butyllithium 2.7 M in toluene, AcroSeal®
Specification Test Results
  • Appearance:
    Yellow to orange Solution (May develop some turbidity or precipitate)
  • Titration Active base:
    19.5 to 20.5 %
  • Free base:
    ≤1.0 %