
4-(Trifluoromethyl)benzylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3300-51-4
:
Formula:C₈H₈F₃N
MDL Number:MFCD00010220
:175.15 g/mol
Boiling Pt:79…82 °C (15 mmHg)
Density:1.229 g/cm³
Flash Pt:75 °C
Storage Temperature:Ambient
EINECS:221-971-9
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4630 to 1.4650 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together