
2-Chloro-4,6-difluoroaniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36556-56-6
:
Formula:C₆H₄ClF₂N
MDL Number:MFCD00142892
:163.55 g/mol
Melting Pt:28…33 °C
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Off-white to grey-green Low melting solid
- Infrared spectrum:Conforms
- Melting point:28 to 33 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together