Order Entry
Czech Republic
ContactUsLinkComponent
Disodium hexachloroplatinate hexahydrate 98%
Health
Corrosive
Disodium hexachloroplatinate hexahydrate 98%
Catalog # ACRO316750010
CAS Number:  
undefined
Health
Corrosive
Disodium hexachloroplatinate hexahydrate 98%
Catalog # ACRO316750010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Orange Crystalline solid
  • Assay:
    ≥97.5 %

Specification Test Results

About this item

 :
Merck Index:12,08768
Density:2,5 g/cm³ (20 °C)
ADR:6.1,III
 :561.87 g/mol
EINECS:240-983-5
 :GHS08
 :GHS05
CAS Number:19583-77-8
Formula:Cl₆Na₂Pt·6H₂O
MDL Number:MFCD00149172