
Tris(pentafluorophenyl)phosphine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1259-35-4
:
Formula:C₁₈F₁₅P
MDL Number:MFCD00079654
:532.15 g/mol
Melting Pt:108…110 °C
EINECS:215-021-2
:GHS07
Specification Test Results
- Appearance:White to beige to light brown Powder or crystals
- Infrared spectrum:Conforms
- Melting point:108 to 119 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together