
3-(Trifluoromethyl)benzylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2740-83-2
:
Formula:C₈H₈F₃N
MDL Number:MFCD00008117
Boiling Pt:93…97 °C (22 mmHg)
Density:1.22 g/cm³
Flash Pt:71 °C
Storage Temperature:Ambient
EINECS:220-367-2
UN:2735
ADR:8,II
:175.15 g/mol
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4620 to 1.4640 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together