
2-(Trifluoromethyl)benzylamine 98%
:3048-01-9
:
:C₈H₈F₃N
:MFCD00010297
:175.15 g/mol
:108 °C (64 mmHg)
:1.249 g/cm³
:69 °C
:Ambient
:221-258-2
:2735
:8,III
:GHS05
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4700 to 1.4720 (20°C, 589 nm)
- Titration with HClO4:≥97.5 %
Frequently Bought Together