
Tris(4-fluorophenyl)phosphine 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18437-78-0
:
Formula:C₁₈H₁₂F₃P
MDL Number:MFCD00013553
:316.26 g/mol
Melting Pt:79…82 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:79 to 83 °C
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together