Order Entry
Czech Republic
ContactUsLinkComponent
Lithocholic acid 98%
Lithocholic acid 98%
Certificates
undefined
Lithocholic acid 98%
Certificates

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    182 to 190 °C
  • HPLC:
    ≥94 %
  • Specific optical rotation:
    +32° to +38° (20 °C, 589 nm) (1.5 % in C2H5OH)

Specification Test Results

Related Information

Frequently Bought Together