ContactUsLinkComponent
Chenodeoxycholic acid 97+%
Irritant
Chenodeoxycholic acid 97+%
Certificates
undefined
Irritant
Chenodeoxycholic acid 97+%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 %
  • Specific optical rotation:
    +9° to +14° (20°C, 589 nm) (c=2, EtOH)
  • Titration with NaOH:
    ≥97.0 % (on dried basis)