ContactUsLinkComponent
n-Butyllithium 2.5 M in hexanes, AcroSeal®
Flammable
Corrosive
Irritant
Health
Environment
n-Butyllithium 2.5 M in hexanes, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Environment
n-Butyllithium 2.5 M in hexanes, AcroSeal®
Certificates
Specification Test Results
  • Appearance:
    Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
  • Titration Active base:
    22.0 to 24.0 %
  • Free base:
    ≤1.2 %