Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Dimethyl-1,1,1-trimethylsilylamine 97%
Corrosive
Flammable
N,N-Dimethyl-1,1,1-trimethylsilylamine 97%
Certificates
undefined
Corrosive
Flammable
N,N-Dimethyl-1,1,1-trimethylsilylamine 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3960 to 1.3980 (20°C, 589 nm)
  • Titration with HClO4:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together