ContactUsLinkComponent
Disodium succinate, anhydrous 99%
Irritant
Disodium succinate, anhydrous 99%
Certificates
undefined
Irritant
Disodium succinate, anhydrous 99%
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.3 % (120°C)
  • Titration with HClO4:
    ≥98.5 %