ContactUsLinkComponent
Tetrakis(triphenylphosphine)palladium(0) 99%
Irritant
Tetrakis(triphenylphosphine)palladium(0) 99%
Certificates
undefined
Irritant
Tetrakis(triphenylphosphine)palladium(0) 99%
Certificates
Specification Test Results
  • Appearance:
    Yellow to yellow-green Powder or crystals
  • Infrared spectrum:
    Conforms
  • Carbon (C):
    73.7 to 76.0 %