ContactUsLinkComponent
Benzyltributylammonium chloride 98+%
Irritant
Benzyltributylammonium chloride 98+%
Certificates
undefined
Irritant
Benzyltributylammonium chloride 98+%
Specification Test Results
  • Appearance:
    White to cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    98.0 to 102.0 %
  • Water:
    ≤1 %
  • HPLC:
    ≥96.0 %