


(2-Methoxyethoxy)methyl chloride 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3970-21-6
:
Formula:ClCH₂OCH₂CH₂OCH₃
MDL Number:MFCD00000888
:124.57 g/mol
Boiling Pt:50…52 °C (13 mmHg)
Density:1.09 g/cm³
Flash Pt:54 °C
EINECS:223-589-8
UN:1992
ADR:3,III
:GHS07
:GHS08
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4280 (20°C, 589 nm)
- GC:≥92 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together