


1,3-Dimethyl-2-imidazolidinone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80-73-9
:
Formula:C₅H₁₀N₂O
MDL Number:MFCD00003188
:114.15 g/mol
Merck Index:13,03275
Boiling Pt:225.5 °C (1013 hPa)
Melting Pt:8.2 °C
Density:1.05 g/cm³
Flash Pt:93 °C
EINECS:201-304-8
ADR:6.1,III
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4710 to 1.4730 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥97.5 %
- Color scale:≤50 APHA
Specification Test Results
Frequently Bought Together