ContactUsLinkComponent
n-Butyllithium 1.6 M in hexanes, AcroSeal®
Irritant
Health
Corrosive
Environment
Flammable
n-Butyllithium 1.6 M in hexanes, AcroSeal®
Certificates
undefined
Irritant
Health
Corrosive
Environment
Flammable
n-Butyllithium 1.6 M in hexanes, AcroSeal®
Specification Test Results
  • Appearance:
    Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
  • Titration Active base:
    14.0 to 16.0 %
  • Free base:
    ≤1 %