
(4-Methoxyphenyl)acetonitrile 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-47-2
:
Formula:C₉H₉NO
MDL Number:MFCD00001919
Boiling Pt:286 °C (1013 hPa)
Melting Pt:8 °C
Density:1.08 g/cm³
Flash Pt:117 °C
EINECS:203-206-0
ADR:6.1,III
:147.18 g/mol
UN:3276
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5290 to 1.5330 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together