1,5-Diphenylcarbazone suitable for colorimetric analysis
:538-62-5
:
:C₁₃H₁₂N₄O
:MFCD00003024
:240.26 g/mol
:12,03380
:440 °C (1013 hPa)
:153…157 °C (dec.)
:Ambient
:208-698-0
- Appearance:Orange to brown Powder
- Infrared spectrum:Conforms
- HPLC:≥35 % (Diphenylcarbazone basis)
- Suitability:(for Hg determination) Passes test
- NMR (1H-NMR):Consistent with structure
Frequently Bought Together