
5-Methoxy-2-methylindole 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1076-74-0
:
Formula:C₁₀H₁₁NO
MDL Number:MFCD00005620
Boiling Pt:145 °C (1,5 -rr)
Melting Pt:85…88 °C
:161.2 g/mol
Storage Temperature:Refrigerator
EINECS:214-066-5
:GHS07
Specification Test Results
- Appearance:White to light beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:85 to 88 °C
- GC:≥99.0 %
- Elemental Analysis:27.71 to 35.27 % (C)
Specification Test Results
Frequently Bought Together