Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxybenzenesulphonyl chloride 99%
Corrosive
4-Methoxybenzenesulphonyl chloride 99%
Certificates
undefined
Corrosive
4-Methoxybenzenesulphonyl chloride 99%

Specification Test Results

  • Appearance:
    White to beige Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    36 to 41 °C
  • GC:
    ≥98.5 %
  • Uniformity coefficient:
    ≤2.00
  • Moisture holding capacity:
    60 to 70 % (H)
  • Total volume capacity:
    ≥2.60 eq/l (H)

Specification Test Results

Related Information

Frequently Bought Together