Order Entry
Czech Republic
ContactUsLinkComponent
Calcium stearate ≥85%
Calcium stearate ≥85%
Catalog # ABCRAB118797.0100
Supplier:  abcr
CAS Number:  
undefined
Calcium stearate ≥85%
Catalog # ABCRAB118797.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :607.03 g/mol
Merck Index:13,01708
Melting Pt:147…149 °C
Density:1.065 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-472-8
CAS Number:1592-23-0
 :66071-81-6
Formula:C₃₆H₇₀CaO₄
MDL Number:MFCD00036390