cis-4-Hydroxy-L-(-)-proline, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:618-27-9
:
Formula:C₅H₉NO₃
MDL Number:MFCD00064319
:131.13 g/mol
Melting Pt:257 °C
:M-Clarity™ quality level = MQ100
EINECS:210-542-1
Frequently Bought Together