L(+)-vinan didraselný hemihydrát ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6100-19-2
:
Formula:KOOC(CHOH)₂COOK·0.5H₂O
MDL Number:MFCD00150136
Merck Index:14,7677
Boiling Pt:200…220 °C
Melting Pt:155 °C (-H₂O)
Density:1,98
Storage Temperature:Ambient
EINECS:213-067-8
Related Information
Frequently Bought Together