Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxy-2,5-dimethylphenylboronic acid 96%
Irritant
4-Methoxy-2,5-dimethylphenylboronic acid 96%
Catalog # APOSOR4108-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methoxy-2,5-dimethylphenylboronic acid 96%
Catalog # APOSOR4108-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:187 - 190 °C
 :180.01 g/mol
 :GHS07
CAS Number:246023-54-1
Formula:C₉H₁₃BO₃
MDL Number:MFCD06801676
Melting Pt:187…190 °C