Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 95%
Toxic
4-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 95%
Catalog # APOSPC99135-1G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
4-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 95%
Catalog # APOSPC99135-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
ADR:6.1,III
 :268.01 g/mol
UN:2811
 :GHS06
CAS Number:2090463-13-9
Formula:C₈H₂BrF₄N
MDL Number:MFCD33404170