
Methyl 5-bromo-3-chloro-2-fluorobenzoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1535973-93-3
:
Formula:C₈H₅BrClFO₂
MDL Number:MFCD24107276
:267.48 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals