Methyl 4-(Bromomethyl)-3-iodobenzoate 95%
Catalog # APOSOR400640-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:354.97 g/mol
CAS Number:229028-10-8
Formula:C₉H₈BrIO₂
MDL Number:MFCD13659352