Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%
Irritant
Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%
Katalogové číslo APOSPC909782-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%
Katalogové číslo APOSPC909782-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.58 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1360934-51-5
Formula:C₈H₅ClF₃NO₂
MDL Number:MFCD22559950